Learn More
Bellco Glass, Inc. Glass Ball Spinner Complete,250mL
SDP
Catalog No. 501945108
Encompass_Preferred
Description: Glass Ball Spinner Comp, 250mL. MADE IN THE USASize: 250mLMaterial: Type 1 Class A Borosilicate GlassFlask Dimensions: 85mm OD x 140mm OAH, two 32mm sidearms offset at 55º, 70mm center neckIncludes two 32mm blue PP screw caps & two 32mm blue PP pouring ringsImpeller Assembly: one 70mm blue PP screw cap, 70mm Teflon cap seal, 70mm blue silicone sealing gasket, soft silicone flex tube, glass ball magnet tube, and magnetSpecially Formed Bottom Indent Prevents Accumulation under impellerGlass ball houses a magnet for gentle agitation of Micro carrier Beads
Catalog No. 50-194-5108 Supplier Bellco Glass, Inc. Supplier No. 196592002
Description: Glass Ball Spinner Comp, 250mL. MADE IN THE USASize: 250mLMaterial: Type 1 Class A Borosilicate GlassFlask Dimensions: 85mm OD x 140mm OAH, two 32mm sidearms offset at 55º, 70mm center neckIncludes two 32mm blue PP screw caps & two 32mm blue PP pouring ringsImpeller Assembly: one 70mm blue PP screw cap, 70mm Teflon cap seal, 70mm blue silicone sealing gasket, soft silicone flex tube, glass ball magnet tube, and magnetSpecially Formed Bottom Indent Prevents Accumulation under impellerGlass ball houses a magnet for gentle agitation of Micro carrier Beads