Learn More
Bellco Glass, Inc. Glass Ball Spinner Complete,6000mL
SDP
Catalog No. 501945112
Encompass_Preferred
Description: Glass Ball Spinner Comp, 6000mL. MADE IN THE USASize: 6000mLMaterial: Type 1 Class A Borosilicate GlassFlask Dimensions: 204mm OD x 410mm OAH, two 45mm sidearms offset at 30º, 100mm center neckIncludes two 45mm blue PP screw caps & two 45mm blue PP pouring ringsImpeller Assembly: one 100mm blue PP screw cap, 100mm Teflon cap seal, 100mm blue silicone sealing gasket, soft silicone flex tube, glass ball magnet tube, and magnetSpecially Formed Bottom Indent Prevents Accumulation under impellerGlass ball houses a magnet for gentle agitation of Micro carrier Beads
Catalog No. 50-194-5112 Supplier Bellco Glass, Inc. Supplier No. 196592060
Description: Glass Ball Spinner Comp, 6000mL. MADE IN THE USASize: 6000mLMaterial: Type 1 Class A Borosilicate GlassFlask Dimensions: 204mm OD x 410mm OAH, two 45mm sidearms offset at 30º, 100mm center neckIncludes two 45mm blue PP screw caps & two 45mm blue PP pouring ringsImpeller Assembly: one 100mm blue PP screw cap, 100mm Teflon cap seal, 100mm blue silicone sealing gasket, soft silicone flex tube, glass ball magnet tube, and magnetSpecially Formed Bottom Indent Prevents Accumulation under impellerGlass ball houses a magnet for gentle agitation of Micro carrier Beads