Learn More
Bellco Glass, Inc. Glass Ball Spinner Only,1000mL No Ctr Cap
SDP
Catalog No. 501945117
Encompass_Preferred
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 1000mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 135mm OD x 240mm Overall Height, 250mm across sidearmsTwo 45mm Sidearms angled 60° offset center100mm Screw Thread Center NeckDimpled BottomGraduations: 100mL to 1000mL, every 100mL – White Ink (Approx Volumes)Supply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings
Catalog No. 50-194-5117 Supplier Bellco Glass, Inc. Supplier No. 196594100
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 1000mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 135mm OD x 240mm Overall Height, 250mm across sidearmsTwo 45mm Sidearms angled 60° offset center100mm Screw Thread Center NeckDimpled BottomGraduations: 100mL to 1000mL, every 100mL – White Ink (Approx Volumes)Supply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings