Learn More
Bellco Glass, Inc. Glass Ball Spinner Only,100mL No Ctr Cap
SDP
Catalog No. 501945114
Encompass_Preferred
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 100mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 63.5mm OD x 135mm Overall Height, 145mm across sidearmsTwo 32mm Sidearms angled 55° offset center70mm Screw Thread Center NeckDimpled BottomGraduations: 20mL to 100mL, every 20mL – White Ink (Approx Volumes)Supply with: Two 32mm PP Blue Screw Caps & PP Blue Pouring Rings
Catalog No. 50-194-5114 Supplier Bellco Glass, Inc. Supplier No. 196594010
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 100mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 63.5mm OD x 135mm Overall Height, 145mm across sidearmsTwo 32mm Sidearms angled 55° offset center70mm Screw Thread Center NeckDimpled BottomGraduations: 20mL to 100mL, every 20mL – White Ink (Approx Volumes)Supply with: Two 32mm PP Blue Screw Caps & PP Blue Pouring Rings