Learn More
Bellco Glass, Inc. Glass Ball Spinner Only,3000mL No Ctr Cap
SDP
Catalog No. 501945118
Encompass_Preferred
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 3000mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 165mm OD x 290mm Overall Height, 268mm across sidearmsTwo 45mm Sidearms angled 53° offset center100mm Screw Thread Center NeckDimpled BottomGraduations: 500mL to 3000mL, every 500mL – White Ink (Approx Volumes)Supply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings
Catalog No. 50-194-5118 Supplier Bellco Glass, Inc. Supplier No. 196594300
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 3000mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 165mm OD x 290mm Overall Height, 268mm across sidearmsTwo 45mm Sidearms angled 53° offset center100mm Screw Thread Center NeckDimpled BottomGraduations: 500mL to 3000mL, every 500mL – White Ink (Approx Volumes)Supply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings