Learn More
Bellco Glass, Inc. Glass Ball Spinner Only,500mL No Ctr Cap
SDP
Catalog No. 501945116
Encompass_Preferred
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 500mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 110mm OD x 170mm Overall Height, 221mm across sidearmsTwo 45mm Sidearms angled 60° offset center100mm Screw Thread Center NeckDimpled BottomGraduations: 50mL to 500mL, every 50mL – White Ink (Approx Volumes)Supply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings
Catalog No. 50-194-5116 Supplier Bellco Glass, Inc. Supplier No. 196594050
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 500mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 110mm OD x 170mm Overall Height, 221mm across sidearmsTwo 45mm Sidearms angled 60° offset center100mm Screw Thread Center NeckDimpled BottomGraduations: 50mL to 500mL, every 50mL – White Ink (Approx Volumes)Supply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings