Learn More
Bellco Glass, Inc. Glass Ball Spinner Only,6000mL No Ctr Cap
SDP
Catalog No. 501945119
Encompass_Preferred
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 6000mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 204mm OD x 410mm Overall Height, 268mm across sidearmsTwo 45mm Sidearms angled 30° offset center100mm Screw Thread Center NeckDimpled BottomGraduations: 1L to 6L, every 1L – White Ink (Approx Volumes)Supply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings
Catalog No. 50-194-5119 Supplier Bellco Glass, Inc. Supplier No. 196594600
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 6000mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 204mm OD x 410mm Overall Height, 268mm across sidearmsTwo 45mm Sidearms angled 30° offset center100mm Screw Thread Center NeckDimpled BottomGraduations: 1L to 6L, every 1L – White Ink (Approx Volumes)Supply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings