Learn More
Next Advance Inc Glass Beads 0.5 mm RNAse Free, 4mL
SDP
Catalog No. NC1629163
Encompass
0.5 mm RNAse free glass beads (density is 2.5 g/cc) 4 mL package.
Catalog No. NC1629163 Supplier Next Advance Inc Supplier No. GB05RNA
0.5 mm RNAse free glass beads (density is 2.5 g/cc) 4 mL package.