Learn More
Electron Microscopy Sciences Glass Beads for Model BS1000 Sterilizer
SDP
Catalog No. 5034051
Encompass_Preferred
Replaced glass beads at least once in three months. Clean the glass beads at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
Model: Model BS1000
Catalog No. 50-340-51 Supplier Electron Microscopy Sciences Supplier No. 6611710
Replaced glass beads at least once in three months. Clean the glass beads at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
Model: Model BS1000