Learn More
Electron Microscopy Sciences Glass Beads for Model BS2000 Sterilizer
SDP
Catalog No. 5034058
Encompass_Preferred
Replaced glass beads at least once in three months. Clean the glass beads at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
Model: Model BS2000
Catalog No. 50-340-58 Supplier Electron Microscopy Sciences Supplier No. 6611720
Replaced glass beads at least once in three months. Clean the glass beads at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
Model: Model BS2000