Learn More
Electron Microscopy Sciences Glass Bead Sterilizer Model BS300, Black
SDP
Catalog No. 501928077
Encompass_Preferred
Gass bead sterilizers are quick and easy to use and are compact and light weight which makes them suitable for table tops or workbenches. They are used to sterilize instruments used in most laboratories and doctors' offices, dentists' offices, clinics, hospitals etc, killing  spores and bacteria within 10 seconds. All units are supplied with one filling of glass beads. Glass beads size range from 1.5 – 2.0 mm diameter.
We suggest that glass beads be replaced at least once in three months and cleaned at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
Model: Model BS300
Color: Black
Catalog No. 50-192-8077 Supplier Electron Microscopy Sciences Supplier No. 66116BK
Gass bead sterilizers are quick and easy to use and are compact and light weight which makes them suitable for table tops or workbenches. They are used to sterilize instruments used in most laboratories and doctors' offices, dentists' offices, clinics, hospitals etc, killing  spores and bacteria within 10 seconds. All units are supplied with one filling of glass beads. Glass beads size range from 1.5 – 2.0 mm diameter.
We suggest that glass beads be replaced at least once in three months and cleaned at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
Model: Model BS300
Color: Black