Learn More
Electron Microscopy Sciences Glass Bead Sterilizer Model BS500
SDP
Catalog No. 5034054
Encompass_Preferred
Gass bead sterilizers are quick and easy to use and are compact and light weight which makes them suitable for table tops or workbenches. They are used to sterilize instruments used in most laboratories and doctors' offices, dentists' offices, clinics, hospitals etc, killing  spores and bacteria within 10 seconds. All units are supplied with one filling of glass beads. Glass beads size range from 1.5 – 2.0 mm diameter.
We suggest that glass beads be replaced at least once in three months and cleaned at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
Model: Model BS500
Catalog No. 50-340-54 Supplier Electron Microscopy Sciences Supplier No. 661165
Gass bead sterilizers are quick and easy to use and are compact and light weight which makes them suitable for table tops or workbenches. They are used to sterilize instruments used in most laboratories and doctors' offices, dentists' offices, clinics, hospitals etc, killing  spores and bacteria within 10 seconds. All units are supplied with one filling of glass beads. Glass beads size range from 1.5 – 2.0 mm diameter.
We suggest that glass beads be replaced at least once in three months and cleaned at least once every 2 weeks. Remove glass beads when the unit is completely cool. Wash with mild soap, rinse and dry thoroughly before re-use.
Model: Model BS500