Learn More
Polysciences, Inc. Glass Beads (150-210m) 250g
SDP
Catalog No. NC0085248
Encompass
Polysciences has a full size range of glass beads that are manufactured from specially selected soda lime glass with a density of ~2.48. 150-210m. Hazards: Harmless-use normal precautions - Handling: Exercise normal care - Storage: Store at room temperature
Catalog No. NC0085248 Supplier Polysciences, Inc. Supplier No. 05483250
Polysciences has a full size range of glass beads that are manufactured from specially selected soda lime glass with a density of ~2.48. 150-210m. Hazards: Harmless-use normal precautions - Handling: Exercise normal care - Storage: Store at room temperature