Learn More
Sigma Aldrich Glass Beads
Catalog No. 184066X500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
500 g
6 x 500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-406-6X500G 6 x 500 g
18-406-500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-406-6X500G Supplier Sigma Aldrich Supplier No. 184066X500G
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Glass beads are generally added to the boiling solutions to prevent bumping. It is also used as supports to various photocatalysts to enhance the adsorption of organic pollutants in photocatalysis.
Specifications
Quantity 6 x 500 g
Synonym Anti-bumping granules