Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Glass Beads, 2 mm
GSA_VA

Catalog No. MP116540441 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116540441 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116540441 Supplier MP Biomedicals™ Supplier No. 116540441
Add to Cart
Add to Cart

Glass Beads, 2 mm are effective at breaking pathogenic yeast polysaccharide capsules and hyphae inside infected tissue.

  • Made of silica glass
  • Sample type: Animal Tissues, Yeast, Fungi
  • Specific Gravity: 2.55

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (English) 0.07 to 0.08 in.
Diameter (Metric) 1.7 to 2.1 mm
For Use With (Equipment) FastPrep-24 5G; FastPrep-24 Homoginizers
For Use With (Application) Sample preparation, sample lysis, nucleic acid isolation and purification, protein isolation, molecular biology, gene expression, microbiological analysis, gene function, gene regulation
Product Type Silica Glass Beads
Quantity 1 kg
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.