Learn More
Sapidyne Instruments Inc GLASS BEADS 500MG TUBE PK/10
SDP
Catalog No. NC2654139
Encompass
  • Glass adsorption beads for increased reliability.
  • Supplied in 10-pack aliquots for convenience.
  • Designed for use in adsorption coating applications.
  • Environmentally conscious alternative to plastic beads.
  • Minimizes concerns related to microplastics.
  • Avoids potential future supply chain disruptions.
  • Engineered for optimal performance with KinExA 4000 instrument.
  • Requires Red Propellers for effective use.
  • Facilitates ease of handling during manufacturing processes.
  • Practical for scientific laboratories focused on molecular interactions.
  • Not considered an environmental concern.
  • No anticipated future supply chain difficulties or bans.
Catalog No. NC2654139 Supplier Sapidyne Instruments Inc Supplier No. 440425
  • Glass adsorption beads for increased reliability.
  • Supplied in 10-pack aliquots for convenience.
  • Designed for use in adsorption coating applications.
  • Environmentally conscious alternative to plastic beads.
  • Minimizes concerns related to microplastics.
  • Avoids potential future supply chain disruptions.
  • Engineered for optimal performance with KinExA 4000 instrument.
  • Requires Red Propellers for effective use.
  • Facilitates ease of handling during manufacturing processes.
  • Practical for scientific laboratories focused on molecular interactions.
  • Not considered an environmental concern.
  • No anticipated future supply chain difficulties or bans.