Learn More
Cole-Parmer Combination pH electrode, Glass, Refillable, Double-Junction, BNC
Catalog No. NC1313277 Shop All Cole-Parmer Products
Encompass_Preferred
Cole-Parmer Combination pH electrode, Glass, Refillable, Double-Junction, BNC, Faster response and increased stability across a pH range of 0 to 14Dome bulb offers better protection from breakageStability in low-ionic strength solutionsOffers resistance to Na+ ion interferenceCompatible with most brands of pH meters (that accept standard BNC connections)
Catalog No. NC1313277 Supplier Cole-Parmer Supplier No. 5900134
Cole-Parmer Combination pH electrode, Glass, Refillable, Double-Junction, BNC, Faster response and increased stability across a pH range of 0 to 14Dome bulb offers better protection from breakageStability in low-ionic strength solutionsOffers resistance to Na+ ion interferenceCompatible with most brands of pH meters (that accept standard BNC connections)