Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp™ Glass Bottle for Solvent Collection
SDP

Catalog No. 11392088
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-392-088 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-392-088 Supplier Velp™ Supplier No. A00000301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Complete glass bottle for use in solvent collection.

  • Collect solvent with ease using this glass collection bottle

Specifications

Capacity (Metric) 2000 mL
Color Clear
Material Glass
Quantity 1 Bottle
Sterility Non-sterile
Closure Included No
Graduated No
Mouth Narrow, Threaded
For Use With (Equipment) SER158/3 Solvent Extractor
Autoclavable Not Autoclavable
Capacity (English) 2 L
Shape Round
Disposable Single-use
Product Type Solvent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.