Learn More
Thomas Scientific Glass Bottom Cell Culture Dish, F 15mm, TC, Sterile
SDP
Catalog No. 501461363
Encompass
Used in confocal microscope, high resolution microscope, Differential Interference Contrast Microscope, Polarized Light Microscope and Phase Contrast Microscope for cell observation Can be used for live cells observation Special bottom design for easy grip Imported optical grade cover glass, cover glass thickness: 0.19-0.22 mm Round cover glass insert for good appearance Medical adhesive to guarantee non-cytotoxicity
Catalog No. 50-146-1363 Supplier Thomas Scientific Supplier No. 1194Z34
Used in confocal microscope, high resolution microscope, Differential Interference Contrast Microscope, Polarized Light Microscope and Phase Contrast Microscope for cell observation Can be used for live cells observation Special bottom design for easy grip Imported optical grade cover glass, cover glass thickness: 0.19-0.22 mm Round cover glass insert for good appearance Medical adhesive to guarantee non-cytotoxicity