Learn More
Bellco Glass, Inc. Glass Bottom Cell Culture Dish,Φ 15mm,TC, sterile 10/pk, 200/cs
SDP
Catalog No. 502330264
Encompass_Preferred
Glass Bottom Cell Culture Dish,Φ 15mm,TC, sterile 10/pk, 200/cs. glass bottom series are applied in confocal microscope, high resolution microscope, differential interference contrast microscope, polarized light microscope and phase contrast microscope for cell observation. • Can be used for live cell observation.• Special bottom design for easy grip.• Round cover glass insert for good appearance.• Medical adhesive glue to guarantee non-cytotoxicity.• Sterilized by E-beam, SAL=10-6.• Made of high clarity, 100% virgin polystyrene and high clarity glass as bottoms.• High quality cover glass of standard thickness.
Catalog No. 50-233-0264 Supplier Bellco Glass, Inc. Supplier No. 451400261
Glass Bottom Cell Culture Dish,Φ 15mm,TC, sterile 10/pk, 200/cs. glass bottom series are applied in confocal microscope, high resolution microscope, differential interference contrast microscope, polarized light microscope and phase contrast microscope for cell observation. • Can be used for live cell observation.• Special bottom design for easy grip.• Round cover glass insert for good appearance.• Medical adhesive glue to guarantee non-cytotoxicity.• Sterilized by E-beam, SAL=10-6.• Made of high clarity, 100% virgin polystyrene and high clarity glass as bottoms.• High quality cover glass of standard thickness.