Learn More
Alkali Scientific Glass Bottom Cell Culture Dish, 35mm Diameter with 15mm Internal Diameter Glass, 10/pk, 200/cs
SDP
Catalog No. 502389550
Encompass_Preferred
The innovative design guarantees a single-plane, flat bottom with a consistent working distance and maximal planaritycombines the conveniance of a standard size 35mm disposable plastic cell culture dish with the optical quality of glass, providing superior high-resolution microscopic images of in-vitro cultivated cultures. Glass bottom features high transparent achromatic borosilicate glass; hydrolytic class 1 (DIN ISO 719) Glass thickness 175 um +/- 15um. Manufactured according to ISO 8255-1; 1986. The subdivided version of the Glass Bottom Cell Culture Dish enables simultaneous multiplex analyzes of different cell lines, varioius stimulations, or diverse transfections. Used in confocal microscope, high resolution microscope, Differential Interference Contrast Microscope, Polarized Light .
Catalog No. 50-238-9550 Supplier Alkali Scientific Supplier No. TC801002
The innovative design guarantees a single-plane, flat bottom with a consistent working distance and maximal planaritycombines the conveniance of a standard size 35mm disposable plastic cell culture dish with the optical quality of glass, providing superior high-resolution microscopic images of in-vitro cultivated cultures. Glass bottom features high transparent achromatic borosilicate glass; hydrolytic class 1 (DIN ISO 719) Glass thickness 175 um +/- 15um. Manufactured according to ISO 8255-1; 1986. The subdivided version of the Glass Bottom Cell Culture Dish enables simultaneous multiplex analyzes of different cell lines, varioius stimulations, or diverse transfections. Used in confocal microscope, high resolution microscope, Differential Interference Contrast Microscope, Polarized Light .