Learn More
Alkali Scientific ASI™ Glass Bottom Cell Culture Dish, 20mm, 10/Pack, 200/Case
SDP
Catalog No. 503028618
Encompass_Preferred
Our glass bottom Cell Culture Dishes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials. We are committed to be your best choice for laboratory and medical consumables. Alkali delivers the one stop solution for its products including tooling making, injection, processing, sterilization, all under one roof. ­­­­­­­­­­­­­­­­­­­­­­­­­­­­­15mm Glass Bottom Cell Culture Dish With ISO9001&13485 certified.  Material:USP VI PolypropyleneHigh quality cover glass of standard thickness 0.16-0.19 mm Can be used for live cell observation. Special bottom design for easy grip. Round cover glass inserts for good appearance. Medical adhesive glue to guarantee non-cytotoxicity. Sterilized by E-beam, SAL=10^-6. Made of high clarity, 100% virgin polystyrene and high clarity glass as bottoms. High quality cover glass of standard thickness.
Catalog No. 50-302-8618 Supplier Alkali Scientific Supplier No. TCN801001
Our glass bottom Cell Culture Dishes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials. We are committed to be your best choice for laboratory and medical consumables. Alkali delivers the one stop solution for its products including tooling making, injection, processing, sterilization, all under one roof. ­­­­­­­­­­­­­­­­­­­­­­­­­­­­­15mm Glass Bottom Cell Culture Dish With ISO9001&13485 certified.  Material:USP VI PolypropyleneHigh quality cover glass of standard thickness 0.16-0.19 mm Can be used for live cell observation. Special bottom design for easy grip. Round cover glass inserts for good appearance. Medical adhesive glue to guarantee non-cytotoxicity. Sterilized by E-beam, SAL=10^-6. Made of high clarity, 100% virgin polystyrene and high clarity glass as bottoms. High quality cover glass of standard thickness.