Learn More
Azzota Corp 35mm Glass Bottom Culture Dish, Ps Material, Thin And Optically Clear Bottom. Round Cover Glass, 1.77 Cm2 Growth Area, Glass Bottom Diameter 15mm. Dia 3.4X1.2 mm, Package Of 10
Catalog No. 507532927
Encompass_Preferred
35mm Glass Bottom Culture Dish, PS Material, thin and optically clear bottom. Round cover glass, 1.77 cm2 Growth Area, Glass Bottom Diameter 15mm. Dia 3.4x1.2 mm, Package of 10
Catalog No. 50-753-2927 Supplier Azzota Corp Supplier No. GBD0115
35mm Glass Bottom Culture Dish, PS Material, thin and optically clear bottom. Round cover glass, 1.77 cm2 Growth Area, Glass Bottom Diameter 15mm. Dia 3.4x1.2 mm, Package of 10