Learn More
Cellvis GLASS BOTTOM CULTURE DISH
Catalog No. NC1603302
Encompass
29 mm glass bottom dish, well size 2 0mm, #1.5 glass-like polymer cover slip(0.175±0.010mm). Designed for high resolution imaging such as confocal microscopy
Catalog No. NC1603302 Supplier Cellvis Supplier No. D29201.5P
29 mm glass bottom dish, well size 2 0mm, #1.5 glass-like polymer cover slip(0.175±0.010mm). Designed for high resolution imaging such as confocal microscopy