Learn More
Electron Microscopy Sciences Sterile Culture Clear Glass Bottom Dishes, 30 mm, Bulk in Sleeve, 50 x 7 mm Dish size
SDP
Supplier:  Electron Microscopy Sciences 7067352
Encompass
Sterile, optical quality, glass bottom dishes provide exceptional imaging quality for applications using inverted microscopes. Glass Bottom Dishes eliminate with using conventional plastic or glass petri dishes and chambers with thick bottoms that require a longer working distance, lower magnification objective. Moreover, plastic dishes cannot be used for DIC or any polarization microscopes due to their inherent birefriengence.
Glass Bottom Dishes are ideal for applications such as: patch clamp recordings where flourescent-tagged (GFP) receptors can be identified for selective study; and many other fluorescent microscopy applications, which require low background scattering of light and reduce intrinsic "auto" fluorescence that occurs when using standard plastic dishes.
Well diameter: 30 mm
Package: Bulk in Sleeve
Dish size: 50 x 7 mm
Catalog No. 50-930-458