Learn More
Advanced Biomatrix 6-well plate glass bottom NON-adherent for BIONOVA X DLP printer
Catalog No. 502255974
Encompass_Preferred
Compatible with the BIONOVA X 3D bioprinter from CELLINK, the bioprinting multi-well plates are the key to high throughput DLP printing. 3D Bioprinted constructs are crosslinked to the bottom of adhesive plates, while in the non-adhesive plates, the constructs will be free-floating.
Catalog No. 50-225-5974 Supplier Advanced Biomatrix Supplier No. D16110025297
Compatible with the BIONOVA X 3D bioprinter from CELLINK, the bioprinting multi-well plates are the key to high throughput DLP printing. 3D Bioprinted constructs are crosslinked to the bottom of adhesive plates, while in the non-adhesive plates, the constructs will be free-floating.