Learn More
Ibidi USA Glass Bottom Dish 35mm - #1.5 Glass Bottom Coverslip
SDP
Catalog No. NC1437332
Encompass
The ibidi product family is comprised of a variety of µ–Slides and µ–Dishes, which have all been designed for high–end microscopic analysis of fixed or living cells. The glass bottom versions of the µ–Slides and µ–Dishes are especially designed for TIRF, super resolution and single molecule applications.The Glass Bottom Dish 35 mm is a standard glass bottom dish for cell culture and high resolution microscopy. Ø 20 mm observation area, #1.5 glass coverslip, uncoated, sterilized, 200 pcs./box
This item is not returnable. View return policy
Catalog No. NC1437332 Supplier Ibidi USA Supplier No. 81218200
The ibidi product family is comprised of a variety of µ–Slides and µ–Dishes, which have all been designed for high–end microscopic analysis of fixed or living cells. The glass bottom versions of the µ–Slides and µ–Dishes are especially designed for TIRF, super resolution and single molecule applications.The Glass Bottom Dish 35 mm is a standard glass bottom dish for cell culture and high resolution microscopy. Ø 20 mm observation area, #1.5 glass coverslip, uncoated, sterilized, 200 pcs./box