Learn More
Sutter Instruments GLASS CAPILARY TUBE 250/PK
SDP
Catalog No. NC9404491
Encompass_Preferred
STANDARD WALL BOROSILICATE TUBING WITHOUT FILAMENT - 1.20mmX0.69mmX10cm. - 250 pieces per package
Catalog No. NC9404491 Supplier Sutter Instruments Supplier No. FGGB1206910
STANDARD WALL BOROSILICATE TUBING WITHOUT FILAMENT - 1.20mmX0.69mmX10cm. - 250 pieces per package