Learn More
World Precision Instrument Standard Glass Capillaries-1B150-6
SDP
Catalog No. 50821817
Encompass
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.
Catalog No. 50-821-817 Supplier World Precision Instrument Supplier No. 1B1506
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.