Learn More
World Precision Instrument Standard Glass Capillaries-1B150-3
SDP
Catalog No. 50821815
Encompass
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.
Catalog No. 50-821-815 Supplier World Precision Instrument Supplier No. 1B1503
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.