Learn More
World Precision Instrument Standard Glass Capillaries-1B150F-4
SDP
Catalog No. 50821819
Encompass
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.
Catalog No. 50-821-819 Supplier World Precision Instrument Supplier No. 1B150F4
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.