Learn More
World Precision Instrument Standard Glass Capillaries-1B100-3
SDP
Catalog No. 50821803
Encompass
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.
Catalog No. 50-821-803 Supplier World Precision Instrument Supplier No. 1B1003
These standard glass capillaries are made with quality, borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.