Learn More
World Precision Instrument Standard Glass Capillaries, 2mm OD-1B200-6
SDP
Catalog No. 50821822
Encompass
These standard glass capillaries are made with quality borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.
Catalog No. 50-821-822 Supplier World Precision Instrument Supplier No. 1B2006
These standard glass capillaries are made with quality borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.