Learn More
World Precision Instrument Standard Glass Capillaries, 2mm OD-1B200F-4
SDP
Catalog No. 50821823
Encompass
These standard glass capillaries are made with quality borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.
Catalog No. 50-821-823 Supplier World Precision Instrument Supplier No. 1B200F4
 Restricted Item
Add to Cart
Add to Cart
These standard glass capillaries are made with quality borosilicate glass and used for applications such as: Microinjection, Electrophysiology, Patch clamp, and Fluid Handling.