Learn More
Perkin Elmer US LLC Glass Cinnabar Spray Chamber with Helix for Optima 2x00/4x00/5x00/7x00 DV/8x00
Catalog No. 502105250
Encompass_Preferred
The Cinnabar spray chamber is a mini-cyclonic spray chamber which gives good sensitivity at very low uptakes. The Cinnabar's design and low volume is ideal for the reduction of memory and matrix effects and very fast washout. It does not include an internal baffle and is not hydrofluoric acid-resistant. Precision is considered very good and purity is considered good. Material: Borosilicate glass Volume: 20 mL Physical reproducibility 1% Matrix effects 5 times lower than with coaxial chambers Low RSD's due to highly accurate construction Positive stop for reproducible nebulizer insertion Application uptake range 20 to 400 µL/min
Catalog No. 50-210-5250 Supplier Perkin Elmer US LLC Supplier No. N0777494
The Cinnabar spray chamber is a mini-cyclonic spray chamber which gives good sensitivity at very low uptakes. The Cinnabar's design and low volume is ideal for the reduction of memory and matrix effects and very fast washout. It does not include an internal baffle and is not hydrofluoric acid-resistant. Precision is considered very good and purity is considered good. Material: Borosilicate glass Volume: 20 mL Physical reproducibility 1% Matrix effects 5 times lower than with coaxial chambers Low RSD's due to highly accurate construction Positive stop for reproducible nebulizer insertion Application uptake range 20 to 400 µL/min