Learn More
MSC Rochester Midland Corporation Glass Cleaner, 55 gal, Liquid Concentrate, Ammonia-Free
Catalog No. 502837566
Encompass
This concentrated glass cleaner is designed for effective cleaning of glass and mirrors. It comes in a large 55 gallon drum, making it suitable for industrial use. The ammonia-free, water-based formula with a mild scent is ideal for various surfaces.
  • Liquid concentrate form.
  • Packaged in a drum container.
  • Ammonia-free solution.
  • Mild scent.
  • 55 gallon container size.
  • Water-based composition.
  • Applicable on porcelain, computer screens, formica, stainless steel, enamel, glass, chrome, ceramic, mirrors, and ceramic tile.
  • Removes dust, bug residues, oil, grease, haze deposits, finger marks, oily soils, dirt, soap scum, water marks, and food stains.
  • Purple color.
  • Water-based, citric acid formula.
  • Features EcoLogo, Biodegradable, Green Seal, and USDA Certified BioBased indicators.
Catalog No. 50-283-7566 Supplier MSC Supplier No. 54709316
This concentrated glass cleaner is designed for effective cleaning of glass and mirrors. It comes in a large 55 gallon drum, making it suitable for industrial use. The ammonia-free, water-based formula with a mild scent is ideal for various surfaces.
  • Liquid concentrate form.
  • Packaged in a drum container.
  • Ammonia-free solution.
  • Mild scent.
  • 55 gallon container size.
  • Water-based composition.
  • Applicable on porcelain, computer screens, formica, stainless steel, enamel, glass, chrome, ceramic, mirrors, and ceramic tile.
  • Removes dust, bug residues, oil, grease, haze deposits, finger marks, oily soils, dirt, soap scum, water marks, and food stains.
  • Purple color.
  • Water-based, citric acid formula.
  • Features EcoLogo, Biodegradable, Green Seal, and USDA Certified BioBased indicators.