Learn More
MSC Windex Ammonia-Free Multi-Surface, Streak-Free Cleaner Refill, 128 oz, 4/Case
Catalog No. 502866545
Encompass
Windex Ammonia-Free Multi-Surface, Streak-Free Cleaner Refill is a liquid cleaning solution designed for effective removal of oil, grease, and dirt across various surfaces. Its multi-surface formulation is suitable for aluminum, stainless steel, glass, plastic, marble, and granite.
  • Ammonia-free formula for safe use on multiple surfaces
  • Streak-free cleaning performance
  • Effectively removes oil, grease, and dirt
  • Suitable for cleaning aluminum, stainless steel, glass, plastic, marble, and granite
  • Comes in a 128 oz refill bottle
  • Features a pleasant scent
Catalog No. 50-286-6545 Supplier MSC Supplier No. 02601938
Windex Ammonia-Free Multi-Surface, Streak-Free Cleaner Refill is a liquid cleaning solution designed for effective removal of oil, grease, and dirt across various surfaces. Its multi-surface formulation is suitable for aluminum, stainless steel, glass, plastic, marble, and granite.
  • Ammonia-free formula for safe use on multiple surfaces
  • Streak-free cleaning performance
  • Effectively removes oil, grease, and dirt
  • Suitable for cleaning aluminum, stainless steel, glass, plastic, marble, and granite
  • Comes in a 128 oz refill bottle
  • Features a pleasant scent