Learn More
3M™ Glass Cleaner & Protector, Ready-To-Use
Catalog No. 17623915 Shop All 3M Products
Change view
Click to view available options
Volume (English):
1 qt.
Catalog No. Volume (English)
17-623-915 1 qt.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-915 Supplier 3M™ Supplier No. 7100020793
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ Glass Cleaner and Protector with Scotchgard™ Protection leaves an invisible layer of protection on glass for easier cleaning after routine use. It reduces fogging on windows and mirrors with repeated use, leaving an attractive shine. Cleans (but may not protect) other surfaces. Not for direct food contact surfaces, vehicles, aquariums or eyewear.
  • Streak-free formula
  • Non-ammoniated formula
  • Works like a heavy-duty glass cleaner without the high levels of solvent or ammonia
  • Keeps glass looking clean longer
  • Dries faster than other glass cleaners tested, saving time
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Cleaner
For Use With (Application) Cleaning
Model 59982
Quantity 12/Cs.
Volume (English) 1 qt.
Volume (Metric) 0.946 L