Learn More
3M™ Glass Cleaner and Protector
Catalog No. 17623921 Shop All 3M Products
Change view
Click to view available options
Volume (Metric):
950 mL
Catalog No. Volume (Metric)
17-623-921 950 mL
1 options
This item is not returnable. View return policy
Catalog No. 17-623-921 Supplier 3M™ Supplier No. 7100038228
Add to Cart
Add to Cart
This item is not returnable. View return policy
Glass Cleaner and Protector
  • Cleans and protects glass for a faster, longer lasting shine
  • Makes light work of fingerprints, lipstick and even permanent marker
  • Reduces fogging on mirrors and other glass surfaces
  • Dries faster than other glass cleaners tested, saving time
  • Non-ammoniated formula with a pleasant smell
  • The first commercial glass cleaner with Scotchgard Protection
  • Dries faster than other glass cleaners tested, saving time
TRUSTED_SUSTAINABILITY
Specifications
Description Ready -To-Use
Volume (Metric) 950 mL