Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Glass, Length x Dia. in mm: 22.2 x 6.35, Length x Dia. in inches: 7/8 x 1/4
SDP
Catalog No. CG200411
Encompass_Preferred
Stir Bar, Magnetic, Glass, Length x Dia. in mm: 22.2 x 6.35, Length x Dia. in inches: 7/8 x 1/4. Borosilicate covered Alnico magnet for use in high temperature reactions.
Catalog No. CG200411 Supplier Chemglass Life Sciences Supplier No. CG200411
Stir Bar, Magnetic, Glass, Length x Dia. in mm: 22.2 x 6.35, Length x Dia. in inches: 7/8 x 1/4. Borosilicate covered Alnico magnet for use in high temperature reactions.