Learn More
BUCHI Glass Column Slurry Filling Set
SDP
Catalog No. 05405469 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
15 to 26 mm I.D. Glass Columns
36 to 49 mm I.D. Glass Columns
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-405-469 36 to 49 mm I.D. Glass Columns
05-405-466 15 to 26 mm I.D. Glass Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-405-469 Supplier BUCHI Supplier No. 044060
Add to Cart
Add to Cart
Slurry filling set for the wet filling and conditioning of glass columns.
  • Example of use: Wet filling and conditioning of glass columns with silica gel particles smaller than 25 μm
Specifications
Type Column Filling
For Use With (Application) Wet Filling or Conditioning of Glass Columns
For Use With (Equipment) 36 to 49 mm I.D. Glass Columns