Learn More
Bellco Glass, Inc. Glass Conical Vial, 0.5mL
SDP
Catalog No. 501945619
Encompass_Preferred
DescriptionThis vial is made of clear borosilicate glass and has concial bottom for complete sample extration. Sample capacity of 0.5mL, 9mm in diameter, and 17mm in height. Vials compatible with our Multi-Tier Plate System. 100 vials per case.Multi-Tier Plate System, is an adaptable, and reusable system, which features a versatile 96 well base plate that can secure vials ranging from 0.5mL to 2.5mL and the ability to isolate individual vials as needed.The ANSI footprint ensures compatibility with all autosamplers and robotic systems.Product DetailsColor: Clear GlassSize: 9x17mmVolume: 0.5mLSilanized: NoBottom: ConicalCase Quantity: 100Country Of Origin: USA
Catalog No. 50-194-5619 Supplier Bellco Glass, Inc. Supplier No. 451001005
DescriptionThis vial is made of clear borosilicate glass and has concial bottom for complete sample extration. Sample capacity of 0.5mL, 9mm in diameter, and 17mm in height. Vials compatible with our Multi-Tier Plate System. 100 vials per case.Multi-Tier Plate System, is an adaptable, and reusable system, which features a versatile 96 well base plate that can secure vials ranging from 0.5mL to 2.5mL and the ability to isolate individual vials as needed.The ANSI footprint ensures compatibility with all autosamplers and robotic systems.Product DetailsColor: Clear GlassSize: 9x17mmVolume: 0.5mLSilanized: NoBottom: ConicalCase Quantity: 100Country Of Origin: USA