Learn More
Research Products International Corp Glass Coplin Staining Jar, 6 per Case
SDP
Catalog No. 50212281
Encompass
Popular jars for staining and processing microscope slides. Fixed slides can be stained and dried without removing from jar. Manufactured from soda lime glass and supplied with a white polypropylene screw cap. Polyethylene lid is vented for use in microwave applications. Internal grooves hold up to ten standard 1'' x 3'' slides.
Catalog No. 50-212-281 Supplier Research Products International Corp Supplier No. 144206
Popular jars for staining and processing microscope slides. Fixed slides can be stained and dried without removing from jar. Manufactured from soda lime glass and supplied with a white polypropylene screw cap. Polyethylene lid is vented for use in microwave applications. Internal grooves hold up to ten standard 1'' x 3'' slides.