Learn More
Research Products International Corp Borosilicate Glass, #1 1/2 Coverslip, 24 x 50mm, 1 Ounce
SDP
Catalog No. 50212500
Encompass
Glass Coverslips are manufactured of high quality borosilicate glass. Feature uniform surface quality, purity and cleanliness. Crystal clear coverslips provide the clearest and most distinct representations required for sensitive analysis and viewing. Ideal for histology, cytology, and microbiology applications. No 1 1/2 coverslip measures 24 x 50 mm. Coverslips are sold by weight and 1 ounce is approximately 50 coverslips per pack.
Catalog No. 50-212-500 Supplier Research Products International Corp Supplier No. 246008
Glass Coverslips are manufactured of high quality borosilicate glass. Feature uniform surface quality, purity and cleanliness. Crystal clear coverslips provide the clearest and most distinct representations required for sensitive analysis and viewing. Ideal for histology, cytology, and microbiology applications. No 1 1/2 coverslip measures 24 x 50 mm. Coverslips are sold by weight and 1 ounce is approximately 50 coverslips per pack.