Learn More
Ibidi USA Glass Coverslips for Chambers, removable (No. 1.5H, 24 mm x 60 mm, Unsterile)
SDP
Supplier:  Ibidi USA 10811
Encompass
Glass Coverslips for 3 Well Chamber, removable, 8 Well Chamber, removable, and 12 Well Chamber, removable. No. 1.5 (170 µm +/- 10 µm) D 263 M Schott glass, 24 mm x 60 mm, unsterile, 100 pcs./box
Catalog No. NC0601315