Learn More
Electron Microscopy Sciences Cylinder w/Hex Base | 100ml| Double Metric Scale
SDP
Catalog No. 501930891
Encompass_Preferred
These borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing and strong, stable, hexagonal bases. A double metric scale with 0.2 mL interval and tolerance. Cylinders come with bumper guards.
Catalog No. 50-193-0891 Supplier Electron Microscopy Sciences Supplier No. 6304404
These borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing and strong, stable, hexagonal bases. A double metric scale with 0.2 mL interval and tolerance. Cylinders come with bumper guards.