Learn More
Electron Microscopy Sciences Glass Cylinder with Double Metric Scale, 10 mL, 0.2 ml
SDP
Catalog No. 501930888
Encompass_Preferred
These glass cylinders with pour spouts feature heavy uniform wall tubing and strong, stable, hexagonal bases. Double metric scale with 0.2 mL interval and tolerance. Cylinders come with bumper guards. Made from borosilicate glass.
Volume: 10 mL
Graduations: 0.2 ml
Catalog No. 50-193-0888 Supplier Electron Microscopy Sciences Supplier No. 6304401
These glass cylinders with pour spouts feature heavy uniform wall tubing and strong, stable, hexagonal bases. Double metric scale with 0.2 mL interval and tolerance. Cylinders come with bumper guards. Made from borosilicate glass.
Volume: 10 mL
Graduations: 0.2 ml