Learn More
Electron Microscopy Sciences Glass Cylinder with Double Metric Scale, 500 mL, 0.2 ml
SDP
Catalog No. 501930893
Encompass_Preferred
These glass cylinders with pour spouts feature heavy uniform wall tubing and strong, stable, hexagonal bases. Double metric scale with 0.2 mL interval and tolerance. Cylinders come with bumper guards. Made from borosilicate glass.
Volume: 500 mL
Graduations: 0.2 ml
Catalog No. 50-193-0893 Supplier Electron Microscopy Sciences Supplier No. 6304406
These glass cylinders with pour spouts feature heavy uniform wall tubing and strong, stable, hexagonal bases. Double metric scale with 0.2 mL interval and tolerance. Cylinders come with bumper guards. Made from borosilicate glass.
Volume: 500 mL
Graduations: 0.2 ml