Learn More
Cole-Parmer Blue Enameled Steel and Glass Dewar Flask with Handle, Lid, and Lid Clamps, 4.5 L
Catalog No. NC1981498 Shop All Cole-Parmer Products
Encompass_Preferred
These economical Dewar flasks, featuring a blue enameled stainless steel outer container with a borosilicate glass inner flask, can safely be used with all liquid gases. Available in four capacity sizes, the Dewar flasks are specially designed for laboratory, hospital, and industrial use. The 1-, 2-, and 4.5-L models are also available with lid clamps.
Catalog No. NC1981498 Supplier Cole-Parmer Supplier No. 0377424
These economical Dewar flasks, featuring a blue enameled stainless steel outer container with a borosilicate glass inner flask, can safely be used with all liquid gases. Available in four capacity sizes, the Dewar flasks are specially designed for laboratory, hospital, and industrial use. The 1-, 2-, and 4.5-L models are also available with lid clamps.