Learn More
Chemglass Life Sciences Glass Dish and Plate, 1.9L
SDP
Catalog No. 501311289
Encompass_Preferred
Glass Dish and Plate, 1.9L. This system offers a contained unit for Northern and Southern blotting. Buffer tray for capillary gel blotting comes with glass plate for setting up transfer. Plate will support the wicking paper and weight. The buffer tray is made from glass. Two sizes of trays and plates are available.
Catalog No. 50-131-1289 Supplier Chemglass Life Sciences Supplier No. CLS1997019
Glass Dish and Plate, 1.9L. This system offers a contained unit for Northern and Southern blotting. Buffer tray for capillary gel blotting comes with glass plate for setting up transfer. Plate will support the wicking paper and weight. The buffer tray is made from glass. Two sizes of trays and plates are available.